missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorophenoxyacetic Acid-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichlorophenoxyacetic acid ring-13C6 |
| CAS | 150907-52-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13C6 C2 H6 Cl2 O3 |
| Quantity | 1 mg |
| Synonym | Acetic acid, (2,4-dichlorophenoxy-13C6)- (9CI), 2,4-D (phenyl-13C6), 2,4-D 13C6, 2,4-D 13C6 (phenyl 13C6) |
| InChI Formula | InChI=1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)/i1+1,2+1,3+1,5+1,6+1,7+1 |
| SMILES | OC(=O)CO[13c]1[13cH][13cH][13c](Cl)[13cH][13c]1Cl |
| IUPAC Name | 2-(4,6-dichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)oxyacetic acid |
| Molecular Weight (g/mol) | 226.99 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?