missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl Chloride, TRC

Product Code. 30383530
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383530 10 g
30374099 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383530 Supplier Toronto Research Chemicals Supplier No. TRCD43232010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7 H5 Cl3
Molecular Weight (g/mol) 195.47
Synonym Benzene, 2,4-dichloro-1-(chloromethyl)-, Toluene, α,2,4-trichloro- (7CI,8CI), 2,4-Dichloro-1-(chloromethyl)benzene, 1-Chloromethyl-2,4-dichlorobenzene, 2,4,α-Trichlorotoluene, 2,4-Dichlorobenzyl chloride, NSC 406892, α,2,4-Trichlorotoluene, alpha,2,4-Trichlorotoluene
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCc1ccc(Cl)cc1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H5Cl3/c8-4-5-1-2-6(9)3-7(5)10/h1-3H,4H2
Formula Weight 193.9457
Chemical Name or Material alpha,2,4-Trichlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.