missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl Alcohol, Mikromol™

Product Code. 17931986
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17931986 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17931986 Supplier Mikromol Supplier No. MM0739.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4-Dichlorobenzyl Alcohol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-dichlorobenzyl alcohol
CAS 1777-82-8
Analyte or Component Names 2,4-dichlorobenzyl alcohol
Shipping Condition Room Temperature
Molecular Formula C7 H6 Cl2 O
Certifications/Compliance ISO 17034
Synonym (2,4-Dichlorophenyl)methanol, 2,4-Dichlorobenzenemethanol, 2,4-Dichlorobenzyl alcohol, Dybenal, Myacide, Myacide SP, NSC 15635, Rapidosept, Oxiconazole Related Compound D
InChI Formula InChI=1S/C7H6Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-3,10H,4H2
SMILES OCc1ccc(Cl)cc1Cl
IUPAC Name (2,4-dichlorophenyl)methanol
Molecular Weight (g/mol) 177.03
Quantity 250 mg
Formula Weight 175.9796 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.