missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzonitrile, 98%

Product Code. 11479352
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479352 5 g 5g
11489352 25 g 25g
11499352 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479352 Supplier Thermo Scientific Alfa Aesar Supplier No. A10113.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6574-98-7 | C7H3Cl2N | 172.008 g/mol

Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

Solubility
Soluble in Methanol. Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-98-7
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016373
InChI Key GRUHREVRSOOQJG-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f
PubChem CID 81050
IUPAC Name 2,4-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)Cl)C#N

Specifications

Melting Point 57°C to 61°C
Quantity 5 g
UN Number UN3439
Beilstein 637738
Solubility Information Soluble in Methanol. Slightly soluble in water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.