missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic Acid, Mikromol™

Product Code. 17961636
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961636 Supplier Mikromol Supplier No. MM0014.05
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4-Dichlorobenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorobenzoic acid
CAS 50-84-0
Analyte or Component Names 2,4-Dichlorobenzoic acid
Shipping Condition Room Temperature
Molecular Formula C7 H4 Cl2 O2
Certifications/Compliance ISO 17025
Synonym 2,4-Dichlorobenzoic Acid, Furosemide Imp. E (EP), 2,4-Dichlorobenzyl Alcohol Impurity E, Furosemide Impurity E, NSC 578, Furosemide EP Impurity E
InChI Formula InChI=1S/C7H4Cl2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)
SMILES O=C(C1=CC=C(Cl)C=C1Cl)O
IUPAC Name 2,4-dichlorobenzoic acid
Molecular Weight (g/mol) 191.01
Quantity 100 mg
Formula Weight 189.9588 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.