missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic acid, 98%

Product Code. 11449513
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449513 1000 g 1000g
11459513 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449513 Supplier Thermo Scientific Alfa Aesar Supplier No. A12372.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-84-0 | C7H4Cl2O2 | 191.007 g/mol

2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2?,1?:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2′,1′:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

Solubility
Soluble in ethanol (100 mg/ml), water (0.48 mg/ml), alcohol, ether, and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

Melting Point 159°C to 164°C
Quantity 1000 g
Beilstein 1868192
Solubility Information Soluble in ethanol (100mg/ml),water (0.48mg/ml),alcohol,ether,and acetone.
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzoic acid
compliance-icons
Product Identifier
  • 2,4-Dichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,4-Dichlorobenzoicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.