missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Artikelnummer. 11403614
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11403614 100 g 100g
11483604 5 g 5g
11493604 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Product Code. 11403614 Supplier Thermo Scientific Alfa Aesar Supplier No. A11259.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16271-33-3
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052712
InChI Key FDTPBIKNYWQLAE-UHFFFAOYSA-N
Synonym 2,4-dichloro-benzenesulfonyl chloride, 2,4-dichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dichloro, 2,4-dichlorobenzenesulphonyl chloride, 2,4-dichlorobenzenesulfonylchloride, 2,4-dichlorophenylsulfonyl chloride, 2,4-dichlorobenzenesulphonylchloride, 2,4-dichlorophenyl chlorosulfone, pubchem5115, acmc-1c9b8
PubChem CID 204419
IUPAC Name 2,4-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 50°C to 58°C
Quantity 100 g
UN Number UN3261
Beilstein 2806192
Sensitivity Moisture sensitive
Formula Weight 245.51
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.