missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroaniline, 99%

Product Code. 11428783
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428783 2500 g 2500g
11408783 100 g 100g
11474090 250 g 250g
11418783 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11428783 Supplier Thermo Scientific Alfa Aesar Supplier No. A10518.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-00-7 | C6H5Cl2N | 162.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-00-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007661
InChI Key KQCMTOWTPBNWDB-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
PubChem CID 11123
ChEBI CHEBI:46635
IUPAC Name 2,4-dichloroaniline
SMILES C1=CC(=C(C=C1Cl)Cl)N

Specifications

Melting Point 59°C to 63°C
Density 1.567
Boiling Point 242°C
Flash Point 149°C (300°F)
Quantity 2500 g
UN Number UN3442
Beilstein 386422
Formula Weight 162.02
Percent Purity 99%
Chemical Name or Material 2,4-Dichloroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.