missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichloroacetophenone, 97%

Product Code. 10207370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207370 250 g Glass bottle 250g
10607061 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10207370 Supplier Thermo Scientific Acros Supplier No. 302432500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2234-16-4 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2234-16-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000581
InChI Key XMCRWEBERCXJCH-UHFFFAOYSA-N
Synonym 2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
PubChem CID 16693
IUPAC Name 1-(2,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 29.0°C to 34.0°C
Color Brown-Yellow to Colorless
Density 1.3200g/mL
Boiling Point 242.0°C
Flash Point 131°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3COCH3
Refractive Index 1.5635
Quantity 250 g
Beilstein 07, II, 219
Specific Gravity 1.32
Solubility Information Solubility in water: ca. 200mg/l in water
Formula Weight 189.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2', 4'-Dichloroacetophenone, 98.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.