missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-7-methylquinoline, TRC

Product Code. 30386804
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386804 100 mg
30369250 1 g
30388162 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386804 Supplier Toronto Research Chemicals Supplier No. TRCD436238100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102878-19-3
Molecular Formula C10H7NCl2
Molecular Weight (g/mol) 212.08
SMILES Cc1ccc2c(Cl)cc(Cl)nc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H7Cl2N/c1-6-2-3-7-8(11)5-10(12)13-9(7)4-6/h2-5H,1H3
Formula Weight 210.9956
Chemical Name or Material 2,4-Dichloro-7-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.