missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-(trifluoromethyl)pyrimidine, 95%, Thermo Scientific™

Product Code. 15299846 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15299846 1 g 1g
15209856 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15299846 Supplier Thermo Scientific Acros Supplier No. 453790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16097-64-6
Molecular Formula C5HCl2F3N2
Molecular Weight (g/mol) 216.98
InChI Key ZTNFYAJHLPMNSN-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-trifluoromethyl pyrimidine, pyrimidine, 2,4-dichloro-6-trifluoromethyl, 2,4-dichloro-6-trifluoromethyl-pyrimidine, pubchem13247, ksc496a7p, 2,6-dichloro-4-trifluoromethyl pyrimidine
PubChem CID 334203
IUPAC Name 2,4-dichloro-6-(trifluoromethyl)pyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)C(F)(F)F

Specifications

Boiling Point 207°C
Flash Point 78°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.98
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-6-(trifluoromethyl)pyrimidine
compliance-icons
Product Identifier
  • 2,4-Dichloro-6-(trifluoromethyl)pyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.