missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyrimidine, 98%, ACROS Organics™

Product Code. 10607142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607142 25 g Glass bottle 25g
10454735 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10607142 Supplier Thermo Scientific Acros Supplier No. 162820250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5424-21-5
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006064
InChI Key BTLKROSJMNFSQZ-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
PubChem CID 79471
IUPAC Name 2,4-dichloro-6-methylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43°C to 47°C
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 163.01
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dichloro-6-methylpyrimidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.