missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylphenol, 98%, Thermo Scientific Chemicals

Product Code. 15461459
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1570-65-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00019988
InChI Key WJQZZLQMLJPKQH-UHFFFAOYSA-N
PubChem CID 15292
IUPAC Name 2,4-dichloro-6-methylphenol
SMILES CC1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 52°C to 56°C
Color Brown
Boiling Point 225°C (literature)
Quantity 1 g
UN Number UN3077
Beilstein 1864865
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
Formula Weight 177.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dichloro-6-methylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'