missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylphenol, 98%

Product Code. p-7055672
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15461459

Brand: Thermo Scientific Alfa Aesar H64312.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1570-65-6 | C7H6Cl2O | 177.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1570-65-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00019988
InChI Key WJQZZLQMLJPKQH-UHFFFAOYSA-N
PubChem CID 15292
IUPAC Name 2,4-dichloro-6-methylphenol
SMILES CC1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 52°C to 56°C
Color Brown
Boiling Point 225°C (literature)
Quantity 1 g
UN Number UN3077
Beilstein 1864865
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
Formula Weight 177.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dichloro-6-methylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichloro-6-methylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4,6-Dichloro-o-cresol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.