missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6,7-dimethoxyquinazoline, TRC

Product Code. 30386941
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386941 5 g 5g
30373580 1 g -
30388305 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386941 Supplier Toronto Research Chemicals Supplier No. TRCD4342305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27631-29-4
Molecular Formula C10 H8 Cl2 N2 O2
Molecular Weight (g/mol) 259.09
Synonym 2,4-Dichloro-6,7-dimethoxyquinazoline, Doxazosin Mesilate Imp. E (EP)
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
SMILES COc1cc2nc(Cl)nc(Cl)c2cc1OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H8Cl2N2O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(12)14-9(5)11/h3-4H,1-2H3
Formula Weight 257.9963
Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.