missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dichloro-6,7-dimethoxyquinazoline, 97%

Product Code. 11392336
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392336 1 g 1g
11302346 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392336 Supplier Alfa Aesar™ Supplier No. L06051.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
2,4-Dichloro-6,7-dimethoxyquinazoline is used as an intermediate in the preparation of potential inhibitors of epidermal growth factor receptor kinases. It is also used as an intermediate in the preparation of terazosin.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27631-29-4
Molecular Formula C10H8Cl2N2O2
Molecular Weight (g/mol) 259.086
MDL Number MFCD00051733
InChI Key DGHKCBSVAZXEPP-UHFFFAOYSA-N
Synonym 2,4-dichloro-6,7-dimethoxy-quinazoline, 2,4-dichloro-6,7-dimethoxy quinazoline, quinazoline, 2,4-dichloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxyquinazolin-4-yl chloride, quinazoline,2,4-dichloro-6,7-dimethoxy, doxazosin impurity e, doxazosin mesilate impurity e ep, pubchem9430, usp doxazosin related compound e usp, acmc-209gyd
PubChem CID 520327
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC

Specifications

Melting Point 175°C to 179°C
Quantity 1 g
Beilstein 190331
Formula Weight 259.09
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.