missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulphamoylbenzoic Acid, Mikromol™

Product Code. 30458401
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458401 4 x 25 mg 100mg
17912226 25 mg 25mg
17972876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458401 Supplier Mikromol Supplier No. MM0014.0300254AMP
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4-Dichloro-5-sulphamoylbenzoic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-5-sulphamoylbenzoic Acid
CAS 2736-23-4
Analyte or Component Names 2,4-Dichloro-5-sulphamoylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H5 Cl2 N O4 S
Certifications/Compliance ISO 9001
Synonym Furosemide Imp. B (EP), 2,4-Dichloro-5-sulphamoylbenzoic Acid, Furosemide Impurity B
InChI Formula InChI=1S/C7H5Cl2NO4S/c8-4-2-5(9)6(15(10,13)14)1-3(4)7(11)12/h1-2H,(H,11,12)(H2,10,13,14)
Solution Type Neat
SMILES NS(=O)(=O)c1cc(C(=O)O)c(Cl)cc1Cl
IUPAC Name 2,4-dichloro-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 270.09
Quantity 4 x 25 mg
Formula Weight 268.9316 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.