missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitrobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11443147
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443147 100 g 100g
11423147 5 g 5g
11433147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443147 Supplier Thermo Scientific Alfa Aesar Supplier No. A16111.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400-70-4
Molecular Formula C7H2Cl2F3NO2
Molecular Weight (g/mol) 259.993
MDL Number MFCD00007073
InChI Key VLVNHMVSVDVAOA-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-nitrobenzotrifluoride, 1,5-dichloro-2-nitro-4-trifluoromethyl benzene, 2,4-dichloro-3,5-nitrobenzotrifluoride, benzene, 1,5-dichloro-2-nitro-4-trifluoromethyl, 2,4-dichloro-5-trifluoromethyl-1-nitrobenzene, pubchem4862, 2,4-dichloro-5-nitro benzotrifluoride, 3-trifluoromethyl-4,6-dichloronitrobenzene, 2,4-dichloro-5-trifluoromethyl nitrobenzene
PubChem CID 621179
IUPAC Name 1,5-dichloro-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])Cl)Cl)C(F)(F)F

Specifications

Melting Point 53°C to 57°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2467459
Formula Weight 260
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.