missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%

Product Code. 11428607
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428607 50 g 50g
11408607 2.5 g 2.5g
11418607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428607 Supplier Thermo Scientific Alfa Aesar Supplier No. A19718.18
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28286-86-4 | C7H5Cl3O2S | 259.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28286-86-4
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.53
MDL Number MFCD00044016
InChI Key DFBKCDYLPLBSJP-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methylbenzene-1-sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulfonylchloride, 2,4-dichloro-5-methylbenzene-1-sulphonyl chloride, 2,4-dichloro-5-methylphenylsulfonyl chloride, acmc-1cpwo, 2,4-dichloro-5-methylphenyl chlorosulfone, 2,4-dichloro-5-methyl-benzenesulfonyl chloride, 2,4-dichloro-5-methylbenzene sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulphonyl chloride, benzenesulfonylchloride,2,4-dichloro-5-methyl
PubChem CID 119914
SMILES CC1=CC(=C(Cl)C=C1Cl)S(Cl)(=O)=O

Specifications

Melting Point 70°C to 73°C
Quantity 50 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 2,4-dichloro-5-methylbenzene-1-sulfonyl chloride
Formula Weight 259.54
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-methylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.