missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
Description
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.
Solubility
Slightly soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.
Solubility
Slightly soluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 66°C to 70°C |
| Quantity | 10 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 179 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,4-Dichloro-5-methoxypyrimidine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction