missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-isopropoxyaniline, 98%

Product Code. 11344985
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344985 1 g 1g
11354985 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344985 Supplier Thermo Scientific Alfa Aesar Supplier No. L18371.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41200-96-8 | C9H11Cl2NO | 220.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41200-96-8
Molecular Formula C9H11Cl2NO
Molecular Weight (g/mol) 220.093
MDL Number MFCD00974411
InChI Key RWRWIICNMOKBIQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-isopropoxyaniline, 2,4-dichloro-5-2-propyloxy aniline, 2,4-dichloro-5-propan-2-yloxy aniline, benzenamine, 2,4-dichloro-5-1-methylethoxy, acmc-20aodr, 2,4-dichloro-5-isopropyloxyaniline, 2,4-dichloro-5-isopropoxyphenylamine, 2,4-dichloro-5-isopropoxy-phenylamine, 2,4-dichloro-5-1-methylethoxy aniline, 2,4-dichloro-5-methylethoxy phenylamine
PubChem CID 170476
IUPAC Name 2,4-dichloro-5-propan-2-yloxyaniline
SMILES CC(C)OC1=C(C=C(C(=C1)N)Cl)Cl

Specifications

Melting Point 40°C to 44°C
Boiling Point 110°C to 112°C (0.2 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 2836827
Formula Weight 220.11
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-isopropoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.