missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-3-nitroquinoline, TRC

Product Code. 30384844
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384844 50 mg -
30388332 10 mg -
30383865 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384844 Supplier Toronto Research Chemicals Supplier No. TRCD43930850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132521-66-5
Molecular Formula C9H4Cl2N2O2
Molecular Weight (g/mol) 243.05
SMILES [O-][N+](=O)c1c(Cl)nc2ccccc2c1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H4Cl2N2O2/c10-7-5-3-1-2-4-6(5)12-9(11)8(7)13(14)15/h1-4H
Chemical Name or Material 2,4-Dichloro-3-nitroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.