missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-(4-nitrophenoxy)benzene, 97%, Thermo Scientific™

Product Code. 10202792
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202792 10 g Amber Glass Bottle 10g
10723063 1 g Amber Glass Bottle 1g
10387244 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10202792 Supplier Thermo Scientific Maybridge Supplier No. RDP00053EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1836-75-5
Molecular Formula C12H7Cl2NO3
Molecular Weight (g/mol) 284.092
MDL Number MFCD00128026
InChI Key XITQUSLLOSKDTB-UHFFFAOYSA-N
Synonym nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
PubChem CID 15787
IUPAC Name 2,4-dichloro-1-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl

Specifications

CAS Min % 97.0
Density 1.3000g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3OC6H4NO2
Quantity 10 g
Formula Weight 284.1
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloro-1-(4-nitrophenoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.