missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromotoluene, 98%, Thermo Scientific™

Product Code. 15480809
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15480809

Brand: Thermo Scientific Alfa Aesar H63869.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31543-75-6
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00052985
InChI Key GHWYNNFPUGEYEM-UHFFFAOYSA-N
Synonym 2,4-dibromotoluene, benzene, 2,4-dibromo-1-methyl, 2,4-bromotoluene, 2,4-dibromo-toluene, pubchem3915, 2,4-dibromo toluene, acmc-1bn9b, ksc497e7f, 2,4-dibromo-1-methyl-benzene, ghwynnfpugeyem-uhfffaoysa
PubChem CID 94421
IUPAC Name 2,4-dibromo-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Br

Specifications

Melting Point -10°C (decomposition)
Density 1.85 g/mL
Boiling Point 243°C (decomposition)
Refractive Index 1.601
Quantity 25 g
Beilstein 2078741
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,4-Dibromotoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.