missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiophene, 90+%

Product Code. 11333167
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333167 25 g 25g
11313167 1 g 1g
11323167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333167 Supplier Thermo Scientific Alfa Aesar Supplier No. L13591.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3140-92-9 | C4H2Br2S | 241.928 g/mol

2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3140-92-9
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00043889
InChI Key WAQFYSJKIRRXLP-UHFFFAOYSA-N
Synonym thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
PubChem CID 2724560
IUPAC Name 2,4-dibromothiophene
SMILES C1=C(SC=C1Br)Br

Specifications

Density 2.16
Boiling Point 215°C
Flash Point >110°C (230°F)
Refractive Index 1.632
Quantity 25 g
UN Number UN2810
Beilstein 107651
Solubility Information It is insoluble in water.
Formula Weight 241.94
Percent Purity ≥90%
Chemical Name or Material 2,4-Dibromothiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.