missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromopropiophenone, 98%, Thermo Scientific™

Product Code. 10096852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096852 25 g Glass bottle 25g
10169042 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10096852 Supplier Thermo Scientific Acros Supplier No. 372810250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 38786-67-3
Molecular Formula C9H8Br2O
Molecular Weight (g/mol) 291.97
MDL Number MFCD00209571
InChI Key GKALOSTUZMFUQB-LURJTMIESA-N
PubChem CID 603907
IUPAC Name 2-bromo-1-(4-bromophenyl)propan-1-one
SMILES C[C@H](Br)C(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84°C to 88°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH(Br)CH3
Quantity 25 g
Synonym 2,4'-dibromopropiophenone, 2-bromo-1-4-bromophenyl propan-1-one, 1-propanone, 2-bromo-1-4-bromophenyl, 2,4-dibromopropiophenone, 2,4'-dibromopropiophenon, acmc-1ago1, alpha,4'-dibromopropiophenone, 1-propanone,2-bromo-1-4-bromophenyl, 2-bromo-1-4-bromophenyl-1-propanone #
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylacetate,ethanol,,methanol
Formula Weight 291.97
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4'-Dibromopropiophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2,4'-Dibromopropiophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.