missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromomesitylene, 98%, Thermo Scientific Chemicals

Catalog Number 11443727
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
6942-99-0
C9H10Br2
277.987
MFCD00013525
CIHJFEWFZJQTFE-UHFFFAOYSA-N
2,4-dibromomesitylene, benzene, 2,4-dibromo-1,3,5-trimethyl, mesitylene, 2,4-dibromo, p-brommesitylbromid, mesitylene,4-dibromo, acmc-1bag9, ksc494c7h, benzene,4-dibromo-1,3,5-trimethyl, #
81368
2,4-dibromo-1,3,5-trimethylbenzene
CC1=CC(=C(C(=C1Br)C)Br)C
This item is not returnable. View return policy
6942-99-0
C9H10Br2
277.987
MFCD00013525
CIHJFEWFZJQTFE-UHFFFAOYSA-N
2,4-dibromomesitylene, benzene, 2,4-dibromo-1,3,5-trimethyl, mesitylene, 2,4-dibromo, p-brommesitylbromid, mesitylene,4-dibromo, acmc-1bag9, ksc494c7h, benzene,4-dibromo-1,3,5-trimethyl, #
81368
2,4-dibromo-1,3,5-trimethylbenzene
CC1=CC(=C(C(=C1Br)C)Br)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Boiling Point 278°C to 279°C
Quantity 100 g
Beilstein 2208246
Formula Weight 278
Percent Purity 98%
Chemical Name or Material 2,4-Dibromomesitylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'