missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromoacetophenone, 98+%

Product Code. 11414664
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414664 100 g 100g
11494654 5 g 5g
11404664 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414664 Supplier Thermo Scientific Alfa Aesar Supplier No. A13687.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-73-0 | C8H6Br2O | 277.943 g/mol

2,4'-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4′-Dibromoacetophenone is used as a reagent for analysis of fatty acids by HPLC as 4-bromophenacyl esters, for use in the protection of phenols and carboxylic acids. It undergoes condensation reactions with aldehydes in the presence of SnCl2 or SmI3 to afford α,β-unsaturated ketones. It is also useful in the esterification of carboxylic acids.

Solubility
Soluble in dimethyl sulfoxide (5 mg/ml), methanol (20 mg/ml), toluene and ethanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-73-0
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00000200
InChI Key FKJSFKCZZIXQIP-UHFFFAOYSA-N
Synonym 2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
PubChem CID 7454
IUPAC Name 2-bromo-1-(4-bromophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Br

Specifications

Melting Point 108°C to 112°C
Odor Pungent
Quantity 100 g
UN Number UN3261
Beilstein 607604
Merck Index 14,1427
Solubility Information Soluble in dimethyl sulfoxide (5mg/ml),methanol (20mg/ml),toluene and ethanol. Insoluble in water.
Formula Weight 277.95
Percent Purity ≥98%
Chemical Name or Material 2,4'-Dibromoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.