missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-fluoroaniline, 97%, Thermo Scientific Chemicals

Product Code. 11487077
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487077 25 g 25g
11477077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487077 Supplier Thermo Scientific Alfa Aesar Supplier No. A14435.14
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141474-37-5 | Molecular Formula: C6H4Br2FN | Molecular Weight: 268.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141474-37-5
Molecular Formula C6H4Br2FN
Molecular Weight (g/mol) 268.91
MDL Number MFCD00042230
InChI Key YJLXEKFYZIBUPJ-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo-6-fluoro, 2,4-dibromo-6-fluorobenzenamine, 2,4-dibromo-6-fluoro-phenylamine, 2,4-dibromo-6-fluoro aniline, pubchem3442, intermediates-zcf02078, ksc494o1h, 2,4-dibromo-6-fluoro-aniline, acmc-1c814, 4,6-dibromo-2-fluorophenylamine
PubChem CID 2736765
IUPAC Name 2,4-dibromo-6-fluoroaniline
SMILES NC1=C(F)C=C(Br)C=C1Br

Specifications

Melting Point 62°C to 64°C
Quantity 25 g
UN Number UN2811
Beilstein 6289292
Formula Weight 268.92
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-6-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.