missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-Fluoro-Benzonitrile, TRC

Product Code. 30385856
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385856 50 mg -
30377252 10 mg 10mg
30387856 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385856 Supplier Toronto Research Chemicals Supplier No. TRCD47239050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393585-66-4
Molecular Formula C7H2NFBr2
Molecular Weight (g/mol) 278.9
IUPAC Name 2,4-dibromo-6-fluorobenzonitrile
SMILES Fc1cc(Br)cc(Br)c1C#N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H2Br2FN/c8-4-1-6(9)5(3-11)7(10)2-4/h1-2H
Formula Weight 276.8538
Chemical Name or Material 2,4-Dibromo-6-Fluoro-Benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.