missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-fluorobenzene, 98%, Thermo Scientific™

Product Code. 10402103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402103 25 mL Glass bottle 25mL
10200643 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402103 Supplier Thermo Scientific Acros Supplier No. 380700250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1435-53-6
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.90
MDL Number MFCD00000283
InChI Key MKHDDTWHDFVYDQ-UHFFFAOYSA-N
Synonym 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
PubChem CID 15034
IUPAC Name 2,4-dibromo-1-fluorobenzene
SMILES FC1=CC=C(Br)C=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 2.0470g/mL
Boiling Point 105°C to 110°C (22.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3F
Refractive Index 1.5820 to 1.584
Quantity 25 mL
Specific Gravity 2.047
Solubility Information Solubility in water: insoluble
Formula Weight 253.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dibromo-1-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.