missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene, 98%, Thermo Scientific Chemicals

Catalog Number 10284950
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
95-80-7
C7H10N2
122.17
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=CC=C(N)C=C1N
This item is not returnable. View return policy
95-80-7
C7H10N2
122.17
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=CC=C(N)C=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Brown to Gray
Boiling Point 283.0°C to 285.0°C
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 25 g
Beilstein 13, 124
Solubility Information Solubility in water: 50g/L (25 c). Other solubilities: soluble in alcohol, ether, chloroform and, benzene
Formula Weight 122.17
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Diaminotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Diaminotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'