missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene, 98%

Product Code. p-100285 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 10284950

Brand: Thermo Scientific Acros 146600250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-80-7 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-80-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Synonym 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
PubChem CID 7261
ChEBI CHEBI:34237
IUPAC Name 4-methylbenzene-1,3-diamine
SMILES CC1=CC=C(N)C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Brown to Gray
Boiling Point 283.0°C to 285.0°C
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 25 g
Beilstein 13, 124
Solubility Information Solubility in water: 50g/L (25 c). Other solubilities: soluble in alcohol, ether, chloroform and, benzene
Formula Weight 122.17
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Diaminotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Diaminotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.