missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminophenol dihydrochloride, 98%

Product Code. p-3757408 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10062630

Brand: Thermo Scientific Acros 212840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-09-7 | C6H10Cl2N2O | 197.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-09-7
Molecular Formula C6H10Cl2N2O
Molecular Weight (g/mol) 197.06
MDL Number MFCD00012979
InChI Key KQEIJFWAXDQUPR-UHFFFAOYSA-N
Synonym 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx
PubChem CID 8715
IUPAC Name 2,4-diaminophenol;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 222°C
Color Beige to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (H2N)2C6H3OH·2HCl
Quantity 25 g
Beilstein 13, 550
Merck Index 14, 2983
Solubility Information Solubility in water: 27.5g/100mL (15°C). Other solubilities: slightly soluble in alcohol and ether
Formula Weight 197.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Diaminophenol dihydrochloride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.