missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diamino-6-hydroxypyrimidine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Diamino-6-hydroxypyrimidine |
| CAS | 56-06-4 |
| Molecular Formula | C4 H6 N4 O |
| Quantity | 5 g |
| Synonym | 4(3H)-Pyrimidinone, 2,6-diamino-, 4(1H)-Pyrimidinone, 2,6-diamino- (8CI,9CI), 2,6-Diamino-4(3H)-pyrimidinone, 2,4-Diamino-6-hydroxypyrimidine, 2,4-Diamino-6-pyrimidinone, 2,6-Diamino-3,4-dihydropyrimidin-4-one, 2,6-Diamino-3H-pyrimidin-4-one, 2,6-Diamino-4(1H)-pyrimidinone, 2,6-Diamino-4-hydroxypyrimidine, 2,6-Diamino-4-pyrimidinol, 2,6-Diaminopyrimidin-4-one, 4-Hydroxypyrimidine-2,6-diamine, 6-Aminoisocytosine, 6-Hydroxy-2,4-diaminopyrimidine, 6-Hydroxy-2,4-pyrimidinediamine, NSC 44914, NSC 680818, NSC 9302 |
| InChI Formula | InChI=1S/C4H6N4O/c5-2-1-3(9)8-4(6)7-2/h1H,(H5,5,6,7,8,9) |
| SMILES | NC1=CC(=O)N=C(N)N1 |
| IUPAC Name | 2,6-diamino-1H-pyrimidin-4-one |
| Molecular Weight (g/mol) | 126.12 |
| Formula Weight | 126.0542 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction