missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxypyrimidine, 96%

Product Code. 10773455 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773455 25 g 25g
10010740 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10773455 Supplier Thermo Scientific Acros Supplier No. 112240250
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-06-4 | C4H6N4O | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-06-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006098
InChI Key SWELIMKTDYHAOY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-hydroxypyrimidine, 2,6-diaminopyrimidin-4-ol, 2,6-diaminopyrimidin-4 1h-one, 2,6-diamino-4-pyrimidinol, 2,6-diamino-4-hydroxypyrimidine, 2,6-diaminopyrimidin-4 3h-one, 2,6-diamino-4-pyrimidinone, 4 1h-pyrimidinone, 2,6-diamino, 2,4-diaminohypoxanthine, 6-aminoisocytosine
PubChem CID 2944
IUPAC Name 2,6-diamino-1H-pyrimidin-4-one
SMILES C1=C(NC(=NC1=O)N)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 270.0°C to 290.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Quantity 25 g
Beilstein 24, 469
Merck Index 15, 2983
Solubility Information Solubility in water: 11.5g/L (20°C).
Formula Weight 126.12
Percent Purity 96%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-hydroxypyrimidine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Diamino-6-hydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.