missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di(tert-butoxy)pyrimidine-5-boronic Acid, TRC

Product Code. 30180232
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180232 100 mg 100mg
30180024 500 mg 500mg
30180422 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180232 Supplier Toronto Research Chemicals Supplier No. TRCD493713100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109299-79-8
Molecular Formula C12H21BN2O4
Molecular Weight (g/mol) 268.12
Synonym [2,4-Di(tert-butoxy)pyrimidin-5-yl]boronic Acid, (2,4-Di-tert-butyloxypyrimidin-5-yl)boronic Acid, B-[2,4-Bis(1,1-dimethylethoxy)-5-pyrimidinyl]boronic Acid;
IUPAC Name (2,4-di-tert-butoxypyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CN=C(N=C1OC(C)(C)C)OC(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H21BN2O4/c1-11(2,3)18-9-8(13(16)17)7-14-10(15-9)19-12(4,5)6/h7,16-17H,1-6H3
Formula Weight 268.16
Chemical Name or Material 2,4-Di(tert-butoxy)pyrimidine-5-boronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.