missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenylthio)-6-fluorobenzonitrile, 97%, Thermo Scientific™

Catalog Number 11470748
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
175204-12-3
C13H7ClFNS
263.714
MFCD00068208
KFJFXRCXNMCNBN-UHFFFAOYSA-N
2-fluoro-6-4-chlorophenylthio benzonitrile, 2-4-chlorophenyl thio-6-fluorobenzonitrile, 2-4-chlorophenyl sulfanyl-6-fluorobenzonitrile, 2-4-chlorophenylthio-6-fluorobenzonitrile, 2-4-chlorophenylthio-6-fluorobenzenecarbonitrile, acmc-20anur, maybridge3_004320, 2-4-chlorophenylsulfanyl-6-fluorobenzonitrile
2737296
2-(4-chlorophenyl)sulfanyl-6-fluorobenzonitrile
C1=CC(=C(C(=C1)SC2=CC=C(C=C2)Cl)C#N)F
This item is not returnable. View return policy
175204-12-3
C13H7ClFNS
263.714
MFCD00068208
KFJFXRCXNMCNBN-UHFFFAOYSA-N
2-fluoro-6-4-chlorophenylthio benzonitrile, 2-4-chlorophenyl thio-6-fluorobenzonitrile, 2-4-chlorophenyl sulfanyl-6-fluorobenzonitrile, 2-4-chlorophenylthio-6-fluorobenzonitrile, 2-4-chlorophenylthio-6-fluorobenzenecarbonitrile, acmc-20anur, maybridge3_004320, 2-4-chlorophenylsulfanyl-6-fluorobenzonitrile
2737296
2-(4-chlorophenyl)sulfanyl-6-fluorobenzonitrile
C1=CC(=C(C(=C1)SC2=CC=C(C=C2)Cl)C#N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 71°C
Quantity 1 g
Formula Weight 263.72
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'