missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)propanoic Acid, TRC

Product Code. 30365963
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365963 50 mg 50mg
30356034 500 mg 500mg
30353016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365963 Supplier Toronto Research Chemicals Supplier No. TRCC38182550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-95-4
Molecular Formula C9H9O2Cl
Molecular Weight (g/mol) 184.62
IUPAC Name 2-(4-chlorophenyl)propanoic acid
SMILES CC(C(=O)O)c1ccc(Cl)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H9ClO2/c1-6(9(11)12)7-2-4-8(10)5-3-7/h2-6H,1H3,(H,11,12)
Formula Weight 184.0291
Chemical Name or Material 2-(4-Chlorophenyl)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.