missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)-3-oxopropanenitrile, 95%, Thermo Scientific™

Product Code. 10792503
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 62538-21-0
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.60
MDL Number MFCD00052669
InChI Key DAEXXSXAEMFPHQ-UHFFFAOYNA-N
Synonym 2-4-chlorophenyl-3-oxopropanenitrile, 2-4-chlorophenyl-2-cyanoacetaldehyde, usaf el-67, alpha-formyl-p-chlorophenylacetonitrile, acetonitrile, 2-p-chlorophenyl-2-formyl, benzeneacetonitrile, 4-chloro-.alpha.-formyl, 3-10-00-03025 beilstein handbook reference, 4-chlorophenylcyanoacetaldehyde, benzeneacetonitrile,4-chloro-a-formyl, 2-formyl-2-4-chlorophenyl acetonitrile
PubChem CID 44090
IUPAC Name 2-(4-chlorophenyl)-3-oxopropanenitrile
SMILES ClC1=CC=C(C=C1)C(C=O)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula ClC6H4CH(CN)CHO
Quantity 10 g
Formula Weight 179.61
Percent Purity 95%
Chemical Name or Material 2-(4-Chlorophenyl)-3-oxopropanenitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'