missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)acetic Acid, TRC

Product Code. 30357029
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357029 10 g -
30355992 1 g 1g
30352007 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357029 Supplier Toronto Research Chemicals Supplier No. TRCC37499510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-88-3
Molecular Formula C8 H7 Cl O3
Molecular Weight (g/mol) 186.59
Synonym Acetic acid, (4-chlorophenoxy)- (9CI), Acetic acid, (p-chlorophenoxy)- (8CI), 2-(4-Chlorophenoxy)acetic acid, (4-Chlorophenoxy)acetic acid, (p-Chlorophenoxy)acetic acid, 2-(p-Chlorophenoxy)acetic acid, 4-CPA, BI 12, HP 55, NSC 8769, NSC 9213, P 10, PCPA, Tomatone, Tomatotone, p-CPA
IUPAC Name 2-(4-chlorophenoxy)acetic acid
SMILES OC(=O)COc1ccc(Cl)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H7ClO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)
Formula Weight 186.0084
Chemical Name or Material 4-Chlorophenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.