missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4’-Chlorobenzoyl)benzoic Acid-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4’-Chlorobenzoyl)benzoic Acid-d4 |
| Molecular Formula | C14 D4 H5 Cl O3 |
| Quantity | 10 mg |
| Synonym | o-(4-Chlorobenzoyl)benzoic Acid-d4, 2-(p-Chlorobenzoyl)benzoic Acid-d4, 4-Chlorobenzophenone-2'-carboxylic Acid-d4, 2-(4-Chlorobenzoyl)benzoic Acid-d4, NSC 7825-d4 |
| InChI Formula | InChI=1S/C14H9ClO3/c15-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)14(17)18/h1-8H,(H,17,18)/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)c2ccc(Cl)cc2)c(C(=O)O)c1[2H] |
| IUPAC Name | 2-(4-chlorobenzoyl)-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 264.697 |
| Formula Weight | 264.049 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?