missing translation for 'onlineSavingsMsg'
Learn More

2-(4-bromophenyl)-5-methyl-1,3,4-oxadiazole, Thermo Scientific™

Product Code. 11366845
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11366845 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11366845 Supplier Thermo Scientific Maybridge Supplier No. CC61710DE
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 41421-03-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7BrN2O
MDL Number MFCD00491620
Quantity 5 g
Synonym 2-4-bromophenyl-5-methyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-bromophenyl-5-methyl, 2-4-bromo-phenyl-5-methyl-1,3,4 oxadiazole, 2-4-bromophenyl-5-methyl-1,3,4 oxadiazole, 2-4-bromo-phenyl-5-methyl-1,3,4 oxadiazo le
InChI Key PQSCYRJMPVYVKU-UHFFFAOYSA-N
SMILES CC1=NN=C(O1)C1=CC=C(Br)C=C1
IUPAC Name 2-(4-bromophenyl)-5-methyl-1,3,4-oxadiazole
Molecular Weight (g/mol) 239.07
PubChem CID 540693
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenyl)-5-methyl-1, 3, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.