missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-2-propanol, 97%, Thermo Scientific Chemicals

Catalog Number 15472339
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2077-19-2
C9H11BrO
215.09
MFCD11870096
AOGYBHJTXLXRSM-UHFFFAOYSA-N
2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
12681920
2-(4-bromophenyl)propan-2-ol
CC(C)(O)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
2077-19-2
C9H11BrO
215.09
MFCD11870096
AOGYBHJTXLXRSM-UHFFFAOYSA-N
2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
12681920
2-(4-bromophenyl)propan-2-ol
CC(C)(O)C1=CC=C(Br)C=C1

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

Solubility
Slightly soluble in water (1.4 g/L) (25°C)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 40°C to 45°C
Color White
Density 1.356 g/mL
Boiling Point 75°C to 85°C (0.1 mmHg)
Refractive Index 1.552
Quantity 250 mg
Beilstein 1935066
Solubility Information Slightly soluble in water (1.4g/L) (25°C)
Formula Weight 215.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)-2-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'