missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)pyridine, ≥95%, Thermo Scientific™

Product Code. 10535194
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10535194 1 g 1g
10525194 250 mg 250mg
10076674 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10535194 Supplier Thermo Scientific Maybridge Supplier No. CC48910DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4783-82-8
Molecular Formula C11H8BrNO
Molecular Weight (g/mol) 250.10
MDL Number MFCD08435900
InChI Key FOKJQGKEVMZAJE-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
PubChem CID 23201935
IUPAC Name 2-(4-bromophenoxy)pyridine
SMILES BrC1=CC=C(OC2=CC=CC=N2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 250.09
Percent Purity 95%
Chemical Name or Material 2-(4-Bromophenoxy)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.