missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)pyridine, ≥95%, Maybridge™

Brand:  Maybridge CC48910DE

Code : 31

Additional Details : CAS Number : 4783-82-8 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10076674

  • £287.00 / 5g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

4783-82-8
250.10
FOKJQGKEVMZAJE-UHFFFAOYSA-N
23201935
BrC1=CC=C(OC2=CC=CC=N2)C=C1
C11H8BrNO
MFCD08435900
2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
2-(4-bromophenoxy)pyridine

Specifications

2-(4-Bromophenoxy)pyridine
4783-82-8
100.0
C11H8BrNO
2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
BrC1=CC=C(OC2=CC=CC=N2)C=C1
250.10
250.09
5g
94.5
95%
MFCD08435900
FOKJQGKEVMZAJE-UHFFFAOYSA-N
2-(4-bromophenoxy)pyridine
23201935
95%
Videos
SDS Documents
Certificates