missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)-N,N-dimethylethylamine, ≥97%, Thermo Scientific™

Product Code. 10708645
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10708645 1 g 1g
10653055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708645 Supplier Thermo Scientific Maybridge Supplier No. CC51510DA
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2474-07-9
Molecular Formula C10H14BrNO
Molecular Weight (g/mol) 244.13
MDL Number MFCD00274471
InChI Key MOVOYJFCKMYLHQ-UHFFFAOYSA-N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Synonym 4-hydroxybutyl acrylate, acrylic acid 4-hydroxybutyl ester, hydroxybutyl acrylate, 1,4-butanediol monoacrylate, 2-propenoic acid, 4-hydroxybutyl ester, unii-4o2a4het1x, 4o2a4het1x, hydroxybutylacrylate, 4-hydroxybutylacrylate, 4-hydroxy-butyl acrylate
SMILES CN(C)CCOC1=CC=C(Br)C=C1
IUPAC Name 4-hydroxybutyl prop-2-enoate
PubChem CID 75588
Formula Weight 244.13
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenoxy)-N, N-dimethylethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.