missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)-N,N-dimethylethylamine, 97%, Thermo Scientific™

Product Code. 15491006
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491006 250 mg 250mg
15440618 1 g 1g
15450618 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491006 Supplier Thermo Scientific Alfa Aesar Supplier No. H54295.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An intermediate for the synthesis of Tamoxifen.

Solubility
Soluble in chloroform, dichloromethane.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2474-07-9
Molecular Formula C10H14BrNO
Molecular Weight (g/mol) 244.13
MDL Number MFCD00274471
InChI Key MOVOYJFCKMYLHQ-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy-n,n-dimethylethanamine, 2-4-bromophenoxy-n,n-dimethylethylamine, 2-4-bromophenoxy ethyl dimethylamine, 4-2-n,n-dimethylethoxy phenylbromide, 4-2-n,n-dimethylaminoethoxy phenylbromide, 2-4-bromo-phenoxy-ethyl-dimethyl-amine, ethanamine, 2-4-bromophenoxy-n,n-dimethyl, n-2-4-bromophenoxy ethyl-n,n-dimethylamine, 2-4-bromo-phenoxy-ethyl-dimethyl, 4-2-dimethylaminoethoxy bromobenzene
PubChem CID 213877
IUPAC Name 2-(4-bromophenoxy)-N,N-dimethylethanamine
SMILES CN(C)CCOC1=CC=C(C=C1)Br

Specifications

Quantity 250 mg
UN Number UN2735
Solubility Information Soluble in chloroform,dichloromethane.
Formula Weight 244.13
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenoxy)-N,N-dimethylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.