missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromomethylphenyl)benzonitrile, Mikromol™

Product Code. 30050257
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050257 25 mg 25mg
30050012 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050257 Supplier Mikromol Supplier No. MM0168.230025
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(4-Bromomethylphenyl)benzonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Bromomethylphenyl)benzonitrile
CAS 114772-54-2
Analyte or Component Names 2-(4-Bromomethylphenyl)benzonitrile
Shipping Condition Room Temperature
Molecular Formula C14 H10 Br N
Certifications/Compliance ISO 17025
Synonym [1,1'-Biphenyl]-2-carbonitrile, 4'-(bromomethyl)-, 4'-(Bromomethyl)[1,1'-biphenyl]-2-carbonitrile, 2-(4-Bromomethylphenyl)benzonitrile, 2-(4'-Bromomethylphenyl)benzonitrile, 2-Cyano-4'-(bromomethyl)biphenyl, 2'-(Cyano)biphenyl-4-ylmethyl bromide, 2'-Cyano-4-(bromomethyl)-1,1'-biphenyl, 4-(2-Cyanophenyl)benzyl bromide, 4-(Bromomethyl)-2'-cyanobiphenyl, 4-Bromomethyl-2'-cyano-1,1'-biphenyl, 4'-(Bromomethyl)-2-cyanobiphenyl, 4'-Bromomethyl-2-cyano-1,1'-biphenyl, 4'-Bromomethylbiphenyl-2-carbonitrile
InChI Formula InChI=1S/C14H10BrN/c15-9-11-5-7-12(8-6-11)14-4-2-1-3-13(14)10-16/h1-8H,9H2
SMILES BrCc1ccc(cc1)c2ccccc2C#N
IUPAC Name 2-[4-(bromomethyl)phenyl]benzonitrile
Molecular Weight (g/mol) 272.14
Quantity 25 mg
Formula Weight 270.9997 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.