missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromo-1-pyrazolyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 15467778
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467778 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467778 Supplier Thermo Scientific Alfa Aesar Supplier No. H61270.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Spezifikation

CAS 1184023-09-3
Molecular Formula C10H6BrN3
Quantity 250 mg
UN Number UN3439
Synonym 2-4-bromopyrazol-1-yl benzonitrile, 2-4-bromo-1h-pyrazol-1-yl benzonitrile, 2-4-bromo-1-pyrazolyl benzonitrile
InChI Key RPTLEHPSCLVUDJ-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C#N)N2C=C(C=N2)Br
IUPAC Name 2-(4-bromopyrazol-1-yl)benzonitrile
Molecular Weight (g/mol) 248.083
PubChem CID 61035135
Formula Weight 248.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(4-Bromo-1-pyrazolyl)benzonitrile
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.